Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK543

Sigma-Aldrich

High Sensitivity Glucose Quantitation Kit

sufficient for 500 fluorometric tests

Synonym(s):

Glucose Detection Kit, High Sensitivity Glucose Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116104
NACRES:
NA.25

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; cancer

shipped in

wet ice

storage temp.

−20°C

General description

Glucose, a monosaccharide, is the most important carbohydrate in biology. It is a source of energy and metabolic intermediate for cell growth. Glucose is one of the main products of photosynthesis and starts cellular respiration in both prokaryotes and eukaryotes. The Glucose Quantitation Assay Kit provides a quick and sensitive method for the measurement of glucose in various biological and food and beverage samples (such as serum, plasma, body fluid, food, growth medium, etc.).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service