Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK489

Sigma-Aldrich

Glucose Uptake Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

Glucose Uptake Assay Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
$1,260.00

$1,260.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
$1,260.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

$1,260.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes; cancer

shipped in

wet ice

storage temp.

−20°C

General description

Glucose uptake has a variety of methods and transporters and depends upon the metabolic demand of the cell type and availability of glucose. There are over ten different facilitated diffusion glucose transporters which transports glucose down its concentration gradient without ATP hydrolysis. In the kidneys, secondary active transport is used to uptake glucose against its concentration gradient to ensure that very little glucose is excreted in urine.

Features and Benefits

  • Linear detection range: 0.1 µM to 5 µM 2-DG6P

Suitability

The kit is suitable for the determination of glucose uptake in whole cells and the evaluation of effects of ligands or drugs on glucose transport.

Principle

The Glucose Uptake Assay Kit is a fluorescent cell-based assay that uses 2-deoxyglucose (2-DG), a widely used glucose analog because it can be taken up by glucose transporters and metabolized by endogenous hexokinase, into 2-deoxyglucose 6-phosphate (2-DG6P). 2-DG6P accumulates intracellularly because it is not a suitable substrate for phosphoglucose isomerase, the next step in glycolysis. The cells are lysed, and excess NADP and glucose 6-phosphate dehydrogenase (G6PDH) is added to metabolize 2-DG6P and generate a molar equivalent amount of NADPH. The NADPH is then measured using a G6PDH recycling reaction to amplify the signal and generate a fluorescent signal measureable at λEx = 530 nm/λEm = 585 nm proportional to the concentration of 2-DG6P.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service