Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3600

Absolute Intensity Calibration Standard for Small Angle X-ray Scattering

NIST® SRM® 3600

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$1,450.00

$1,450.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$1,450.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,450.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

form

solid

manufacturer/tradename

NIST®

technique(s)

diffraction/scattering: suitable

application(s)

general analytical

General description

Each unit of SRM 3600 contains a single glassy carbon coupon specimen measuring approximately 10 mm x 10 mm x 1 mm in dimension. For more information, please refer to the SDS and COA.
SRM 3600_cert
SRM 3600_SDS

Application

This Standard Reference Material (SRM) is intended for the scattering intensity calibration of small-angle X-ray scattering (SAXS) instruments. SAXS measurements characterize the microstructure and nanostructure of heterogeneous material systems: specifically, the size distributions of microscale and nanoscale features and, with scattering intensity calibration, their volume fractions or number concentrations, and their surface areas.

Features and Benefits

  • NIST certified values for absolute X-ray scattering intensity, are expressed as the X-ray differential scattering cross-section, dΣ/dΩ, per unit sample volume of glassy carbon, with respect to the magnitude of the scattering vector, Q .
  • Expiration details, instructions for use, storage, and handling are provided on the NIST certificate.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service