Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3254

Camellia sinensis (green tea) leaves

NIST® SRM® 3254

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 3 g

manufacturer/tradename

NIST®

format

matrix material

General description

One unit of SRM 3254 comprises five packets,each containing roughly 3 grams of leaf powder.



SRM 3254_cert

SRM 3254_SDS

Application

Camellia sinensis (green tea) leaves standard reference material (SRM) is primarily intended to validate analytical methods for determining catechins, xanthine,and elemental composition in green tea leaves and similar matrices. Moreover,SRM 3254 can serve as a means of quality assurance for assigning values to in-house control materials. This SRM has also been analyzed for its DNA sequence.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction value of catechins, xanthines, and elements reported on a dry mass basis.
  • The reference values are reported as of mass fraction values for catechin, epicatechin, gallic acid, theanine, andother elements, all reported on a dry-mass basis.
  • The certified value is metrological traceability to the SI derived unit for mass fraction (expressed as milligrams per gram).
  • The NIST certificate is provided with expiration certificate, storage, handling,use, and maintenance instructions.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: (–)-Epicatechin, (–)-Epicatechin gallate, (–)-Epigallocatechin, (–)-Epigallocatechin gallate, (–)-Gallocatechin gallate, Caffeine, Theobromine, (+)-Catechin, (–)-Gallocatechin, Gallic acid, L-Theanine

Matrix Group: Botanicals

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service