Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST675

Line position, Mica

NIST® SRM® 675, for X-ray powder diffraction

Synonym(s):

Fluorphlogopite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg3K[AlF2O(SiO3)3]
CAS Number:
Molecular Weight:
421.24
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

powder

packaging

pkg of 7.5 g

manufacturer/tradename

NIST®

technique(s)

diffraction/scattering: suitable

General description

For more information,please refer to the COA and SDS.

SRM 675_cert

SRM 675_SDS

Application

The Standard Reference Material (SRM) Line position, Mica is intendedfor use as a calibration standard, either externally or internally, for low 2 (large d-spacing) in powder diffractometry.

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service