Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3152A

Sodium (Na) standard solution

NIST® SRM® 3152a

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$2,160.00

$2,160.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
$2,160.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$2,160.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 50 mL

manufacturer/tradename

NIST®

format

single component solution

General description

The SRM 3152A unit consists of 50 mL of an aqueous sodium solution in a high-density polyethylene bottle, sealed in an aluminized bag. The solution was prepared gravimetrically to contain a known mass fraction of sodium in nitric acid at a mass fraction of 5 %.

SRM 3152A _SDS

SRM 3152A _CoA

Application

Sodium (Na) standard solution standard reference material (SRM) is intended for use as a primary calibration standard for the quantitative determination of sodium.

Features and Benefits

  • Available with a certified mass fraction value for sodium.
  •   Certified value is based on; 
1. Gravimetric preparation using high-purity sodium chloride. 

2. Inductively coupled plasma optical emission spectrometry (ICP-OES) calibrated using four primary standards independently prepared from high-purity sodium chloride. 
  • Certified value is metrologically traceable to the International System of Units (SI) derived unit for mass fraction, expressed as milligrams per gram.   
  • The NIST certificate is provided with storage, handling, and use instructions.

Other Notes

Certified for the analytes listed below.
Sodium (Na)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service