NIST3109A
Calcium (Ca) standard solution
NIST® SRM® 3109a
Select a Size
Select a Size
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 10 mL
manufacturer/tradename
NIST®
format
single component solution
Related Categories
General description
SRM 3109A_cert
SRM 3109A_SDS
Other Notes
Calcium (Ca)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service