Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3122

Hafnium (Hf) standard solution

NIST® SRM® 3122

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 50 mL

manufacturer/tradename

NIST®

format

single component solution

General description

This Standard Reference Material (SRM) is intended for use as a primary calibration standard for the quantitative determination of hafnium. A unit of SRM 3122 consists of 50 mL of an acidified aqueous solution prepared gravimetrically to contain a known mass fraction of hafnium in a high-density polyethylene bottle sealed in an aluminized bag. The solution contains approximately 8 % nitric acid (volume fraction) and approximately 1 % hydrofluoric acid (volume fraction), equivalent to amount-of-substance concentrations (molarities) of 1.3 mol/L and 0.3 mol/L, respectively. For more information, please refer to the SDS and COA.

SRM 3122_cert

SRM 3122_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Hafnium (Hf)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service