Skip to Content
MilliporeSigma
All Photos(1)

Documents

42992

Supelco

Magnesium Standard for AAS

TraceCERT®, 1 g/L Mg in nitric acid (nominal concentration)

Synonym(s):

Magnesium nitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(NO3)2 · 6 H2O
CAS Number:
Molecular Weight:
256.41
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1 g/L Mg in nitric acid (nominal concentration)

technique(s)

AAS: suitable

application(s)

industrial qc
pharmaceutical

format

single component solution

InChI

1S/Mg.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;

InChI key

MFUVDXOKPBAHMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Magnesium standard solution has been used in the determination of magnesium ions, which is present in sea -water, using atomic absorption spectrophotometry (AAS).

Analysis Note

1 g/L Mg in 2% nitric acid, prepared with high purity Mg(NO3)2, HNO3 and water

Other Notes

For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of calcium and magnesium in sea-water using a dynamically coated porous graphitic carbon column with a selective metallochromic ligand as a component of the mobile phase
Paull B, et al.
Analytical Communications, 33, 193-196 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service