Merck
Zinc in attention-deficit/hyperactivity disorder.
Journal of child and adolescent psychopharmacology (2005-09-30)
L Eugene Arnold, Robert A DiSilvestro
RESUMO
The aim of this study was to review the published evidence for a role of zinc nutrition in attention-deficit/hyperactivity disorder (ADHD). A computer literature search was supplemented by the authors' knowledge. Numerous controlled studies report cross-sectional evidence of lower zinc tissue levels (serum, red cells, hair, urine, nails) in children who have ADHD, compared to normal controls and population norms. A few studies show correlations of zinc level with either clinical severity or a change thereof in response to stimulant or chemical challenge. Two placebo-controlled trials--one of zinc monotherapy, the other of zinc supplementation of methylphenidate--reported significant benefit. However, diagnostic procedures and sample representativeness were often not clear, and most such reports have come from countries and cultures with different diets and/or socioeconomic realities than are found in the United States (only one American sample in nine published reports). In particular, both positive clinical trials of zinc supplementation came from the Mid-East (Turkey and Iran), an area with suspected endemic zinc deficiency. The largest of these trials used zinc doses above the recommended upper tolerable limit and had a 2 in 3 dropout rate. It is not clear how well the accumulating evidence for a possible role of zinc in ADHD applies to middle-class American children. However, the evidence appears strong enough to warrant further controlled study in well-diagnosed samples representative of the socioeconomic spectrum. Hypothesis-testing clinical trials are needed of this potential treatment that, if found effective, might become a relatively safe, cheap substitute for, or adjunct to, current treatments in some patients. At present, it should remain an investigational treatment.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular