Documentos Principais
57695
Rheodyne® Model 9010 Injector
About This Item
Código UNSPSC:
41115854
Características
manual
number of ports 6
positions 2
Parâmetros
5000 psi max. pressure
Descrição geral
This injector is similar in function to the Model 7010 stainless steel injector, but is constructed of inert PEEK and Tefzel materials for use in the pH range 0-14. It can be used for HPLC, soft gel chromatography, or ion chromatography.
A position-sensing switch closes in the INJECT position and stays closed until the handle is returned to the LOAD position.
Supplied with a 20 μL sample loop; order additional loops separately.
Produtos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informações legais
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)