Documentos
26705
Hamilton® chaney adapter
for use with Hamilton 800/1800 series (5-500 μL)
About This Item
Código UNSPSC:
41122001
Materiais
aluminum
Características
For consistent, reproducible injections
Prevents syringe plunger bending
embalagem
pkg of 1 ea
fabricante/nome comercial
Hamilton 32146
compatibilidade
for use with Hamilton 800/1800 series (5-500 μL)
Descrição geral
Recommended for sample reproducibility.
Informações legais
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificados de análise (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atendimento ao cliente