Documentos Principais
Z740479
MICROGARD 2000 coveralls for protection from infective agent
size 2XL (boot ends with anti-slip soles)
About This Item
Código UNSPSC:
46181503
NACRES:
NB.35
tamanho
2XL (boot ends with anti-slip soles)
fabricante/nome comercial
Arco 1762307
Descrição geral
MICROGARD 2000 fully elasticated coverall with hood , setting the standard in Type 5/6 performance and protection, offers protection from infective agents, pesticides, particulates, fibres and liquid chemical spray.
Features:
  • 3-piece hood
  • 2-way front zipper with re-sealable storm flap
  • Finger loops
  • Elasticated hood, waist, wrists and ankles
Approved to:
EN ISO 13982-1 Type 5 Particulate protection
EN13034 Type 6 Reduced spray
EN1073-2 Barrier to radioactive particulates - Class 2
EN14126 Barrier to infective agents
EN1149-5 Anti-static
Informações legais
Microgard is a trademark of Microgard Limited
Escolha uma das versões mais recentes:
Certificados de análise (COA)