Documentos Principais
Z710857
Silicone membrane liner for Tunair shake flask
For 300 mL flask
Selecione um tamanho
5 EA
R$ 1.141,00
R$ 1.141,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
5 EA
R$ 1.141,00
About This Item
Código UNSPSC:
41122104
NACRES:
NB.22
R$ 1.141,00
Check Cart for Availability
Materiais
silicon
esterilidade
non-sterile
embalagem
case of 5
fabricante/nome comercial
IBI SS-3016
Descrição geral
Silicone membrane liner for Tunair shake flask consists of five 0.22 m silicone membrane cap linings. The filter lining in the cap assembly is replaced by tweaking the flanges of the inner-closure shell until they separate loose. Then it is pulled apart to remove the used lining. The lining can then be replaced by sandwiching it between two parts of the cap and the cap is snapped back together. While reassembling, ensure the flanges from the inner piece fits into the mated grooves in the outer piece. It is required for the cap assembly to stay intact while using.
Características e benefícios
  • Long-lasting unless punctured
  • Rinse with water only if visibly soiled.
  • Recommends autoclaving cap along with the filter.
Informações legais
Tunair is a trademark of IBI Scientific
Escolha uma das versões mais recentes:
Certificados de análise (COA)