Documentos Principais
Z328782
BRAND® magnetic stirring bar, PTFE, cylindrical
L 3.5 mm, bar diam. 3.5 mm
Selecione um tamanho
10 EA
R$ 638,00
R$ 638,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
10 EA
R$ 638,00
About This Item
Código UNSPSC:
41103806
NACRES:
NB.72
R$ 638,00
Check Cart for Availability
Materiais
PTFE-coated
cylindrical
fabricante/nome comercial
BRAND 137101
BRAND
Comprimento
3.5 mm
diâmetro da barra
3.5 mm
Descrição geral
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Características e benefícios
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Outras notas
BR137101 (India)
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)