Documentos Principais
P1049
Sigma-Aldrich
Microcapillary tube, calibrated
capacity 50 μL
Selecione um tamanho
1 PKG
R$ 754,00
R$ 754,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 PKG
R$ 754,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
R$ 754,00
Check Cart for Availability
Materiais
glass
embalagem
pack of 250 ea
fabricante/nome comercial
Drummond Scientific 2-000-050
faixa de medição
accuracy: ±0.5%
capacidade
50 μL
cor
color-coded green
Descrição geral
  • Drummond Calibrated Micropipettes provide accuracy and convenience for a wide variety of liquid transfer applications.
  • Made of high quality glass.
  • Fire polished micropipettes are imprinted with volume and a color-code bar for easy identification.
  • Calibrated "To Deliver."
High quality Drummond micropipettes are fire polished, imprinted with volume and a color-code bar for easy identification. Calibrated "To Deliver."
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Questions
  1. Hello, would it be possible to know the diameter of these capillaries please?
    1 answer
    1. This product has an outer diameter of 1.372 mm or 0.0540 in. and an inner diameter of 0.9357 mm or 0.03684 in.