Documentos Principais
HS120246
Wide mouth wash bottle
capacity 500 mL, red PE-LD bottle
About This Item
Código UNSPSC:
41103209
NACRES:
NB.43
Materiais
polypropylene cap
red PE-LD bottle
Características
label no label
vented
with cap
embalagem
pack of 6 ea
fabricante/nome comercial
Heathrow Scientific HS120246
capacidade
500 mL
Descrição geral
Color-coded tops for easy identification
The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
  • Wide bottle mouth makes filling easier
  • Color-coding for user, application or location identification
  • Bottle is made from LDPE, with a PP closure
  • 500 mL bottle size,
  • Supplied in packs of 6
Escolha uma das versões mais recentes:
Certificados de análise (COA)