Documentos
Z530999
Sigma-Aldrich
Aldrich® Liebig condenser
Overall H 300 mm, joint: ST/NS 24/40
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
Junta
joint: ST/NS24/40
Total A
300 mm
Descrição geral
Condensers have a standard taper female joint and a lower male drip tip-joint.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificados de análise (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atendimento ao cliente