Documentos Principais
MDE6507FS1
Millipore
Filtro de profundidade de cápsula Millistak+®
DE65, filtration area 0.77m², Process Scale
About This Item
Código UNSPSC:
23151806
Preço e disponibilidade não estão disponíveis no momento.
Nome do produto
Filtro de profundidade de cápsula Millistak+®, DE65 (media series), cellulose membrane (and inorganic filter aid), effective filtration area 0.77 m2
Materiais
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane (and inorganic filter aid)
polypropylene housing (glass-filled)
Nível de qualidade
Compatibilidade de esterilização
autoclavable compatible
área efetiva de filtração
0.77 m2
conector
seal (flat seal)
código do cartucho
DE65 (media series)
Descrição geral
The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MDE6507FS1 comes in a POD format.
This DE65 media series delivers optimal performance through a gradient density matri, ideal for primary or secondary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Características e benefícios
  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces
Informações legais
MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany
O que é o Programa Emprove®?
O programa Emprove® é um sistema que fornece documentação completa e aprofundada sobre os nossos filtros e componentes descartáveis, matérias-primas para a indústria farmacêutica e materiais de partida.
Material Qualification Dossier
Gratuito - Faça login para baixar
Saiba mais sobre os benefícios uma assinatura do Programa Emprove® pode fornecer
Por que esses documentos não estão disponíveis?
Os dossiês Emprove® contêm informações destinadas a auxiliar no processo de registro e nas atividades de avaliação de risco dos produtores de fármacos e contêm informações confidenciais. Por favor, entre em contato conosco se desejar ter acesso.