Documentos Principais
Z742609
Sigma-Aldrich
Photo KitAlysis 24-White LED Array
About This Item
Código UNSPSC:
12352300
NACRES:
NA.22
Descrição geral
White LED Array for Photo KitAlysis high-throughput screening platform. For use with Photo KitAlysis Starter Kit (Z742612). User guide is provided in the below hyperlink.
Photo KitAlysis Operating Instructions
Features:
  • Designed and tested by synthetic chemists.
  • Controller provides repeatable milliamp selection for photon intensity (sold seperately)
  • 0-30 mA variable LED output
  • Non-magnetic LED base
  • Chemically resistant LED cover
  • PTFE coated cabling
Photo Kitalysis Starter Kit required for operation (sold separately). Best when used with KitAlysis Benchtop Inertion Box (sold separately).
Informações legais
KitAlysis is a trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Artigos
PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.
Protocolos
Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit