Documentos Principais
Z678740
FORTUNA® Polyfix® bottle top dispenser with PTFE coated piston
dispensing volume 2-10 mL
Selecione um tamanho
1 EA
R$ 1.557,00
R$ 1.557,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 1.557,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
R$ 1.557,00
Check Cart for Availability
fabricante/nome comercial
Fortuna Graf 101.071-38
dispensing volume measuring range
2-10 mL
volume
2-10 mL
conector
GL 40
GL 45
Categorias relacionadas
Descrição geral
For dispensing of weak acids and alkaline solutions
Features:
  • Visible dispensing process by the use of a dispensing cylinder made of glass, coated with PP-protection against breakage
  • Coated PTFE-piston
  • Ergonomic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
  • FIX adapter made of PP
  • Valve block made of PP, no metal springs in the valve - no corrosion
Supplied as FORTUNA Polyfix dispenser, 2 PP adapters GL40 + GL45 & 1 PTFE intake tube
Informações legais
FORTUNA is a registered trademark of Poulten & Graf Ltd.
POLYFIX is a registered trademark of Poulten & Graf Ltd.
Escolha uma das versões mais recentes:
Certificados de análise (COA)