Documentos Principais
Z675946
Tubby® containers
Original
Selecione um tamanho
5 EA
R$ 391,00
R$ 391,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
5 EA
R$ 391,00
About This Item
Código UNSPSC:
41123300
NACRES:
NB.51
R$ 391,00
Check Cart for Availability
Materiais
polypropylene
Características
Original
fabricante/nome comercial
Heathrow Scientific HS23453
C × L × P
330 mm × 200 mm × 115 mm
Descrição geral
5 tubs/lids + 15 dividers
These sturdy tubs with lids and dividers will help you organize your laboratory. Using the removable dividers you can create as big or as small a compartment as you need. Perfect for keeping tubes, tips, pipettes, gloves, and other small items organized and out of the way. The lid snaps in place to secure the contents of the Tubby container. All components are constructed of high-impact polypropylene.
All Tubby containers are proportionally sized to utilize space efficiently when stacked. Tubby Junior is shallow for smaller items. The original Tubby has the same footprint but is twice as deep. Tubby Senior is the size of two original Tubby containers, for your largest items.
Informações legais
Tubby is a registered trademark of Heathrow Scientific
Escolha uma das versões mais recentes:
Certificados de análise (COA)