Documentos Principais
Z505056
Ace O-rings, FETFE®
I.D. 21.8 mm, wall size 3.5 mm
Selecione um tamanho
1 PKG
R$ 258,00
R$ 258,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 PKG
R$ 258,00
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
R$ 258,00
Check Cart for Availability
embalagem
pkg of 6 ea
fabricante/nome comercial
Ace Glass 7855734
D.I.
21.8 mm
tamanho da parede
3.5 mm
Informações legais
FETFE is a registered trademark of Ace Glass, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 9
1 of 9
Ace O-rings, FETFE® I.D. 12.4 mm, wall size 1.78 mm
Z504742
Ace O-rings, FETFE®
Ace pressure tube bushing type, Front seal, capacity ~38 mL, L × O.D. 20.3 cm × 25.4 mm
Z181110
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity ~9 mL, L × O.D. 10.2 cm × 19 mm
Z564567
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity ~21 mL, L × O.D. 20.3 cm × 19 mm
Z564621
Ace pressure tube
Ace pressure tube bushing type, Back seal, capacity ~35 mL, L × O.D. 17.8 cm × 25.4 mm
Z181072
Ace pressure tube
Ace pressure tube bushing type, Back seal, capacity ~15 mL, L × O.D. 10.2 cm × 25.4 mm
Z181064
Ace pressure tube
Ace pressure tube bushing type, Front seal, capacity ~15 mL, L × O.D. 10.2 cm × 25.4 mm
Z181099
Ace pressure tube