Documentos Principais
Z420085
STAT-O-LOK quick-disconnect
male, for tubing i.d., 0.25
Selecione um tamanho
1 EA
R$ 643,00
R$ 643,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 643,00
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
R$ 643,00
Check Cart for Availability
Características
for tubing i.d. 0.25
fabricante/nome comercial
Bel-Art
Descrição geral
  • Chrome-plated brass connectors with slide locks are leak-free
  • Automatic shut-off when disconnected
  • Hose barb ends accept flexible tubing
  • Operate at temperatures to 82°C (180°F), pressures to 250psi, vacuum to 30μm
  • Delrin® valves provide dependable operation
  • Ideal for liquid and gas connectors in biomedical instrumentation and general industrial applications
Informações legais
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
STAT-O-LOK is a trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)