Documentos Principais
Z314986
Hirschmann® pipetus®-Standard pipette controller
with pump
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
entrada CA/CC
230 V AC
Materiais
plastic
silicone
Características
CE compliant
fabricante/nome comercial
Hirschmann 9903000
disponibilidade
not available in USA and Canada
volume
0.1-200 mL
Descrição geral
Similar to the Pipetus-Akku, but with separate pump unit. The separate pumps avoids any contamination and lowers ignition risk. There are no electronic parts within the controller.
Supplied with tubing, hand-held controller, pipette holder, spare filters, spare safety valve and instruction manual.
UK & European version supplied with 2-pin plug and 3-pin adapter.
Informações legais
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Pipetus is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Escolha uma das versões mais recentes:
Certificados de análise (COA)