Documentos Principais
Z285455
Sigma-Aldrich
Aldrich® straight vacuum-distilling adapter
joint: SJ 35/25
Selecione um tamanho
1 EA
R$ 497,00
R$ 497,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 497,00
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
R$ 497,00
Check Cart for Availability
Junta
joint: SJ35/25
Descrição geral
With side hose connection and drip-tip. Improves efficiency of evaporation of solvents under reduced pressure when placed between the condenser of a rotary evaporator and a chilled receiving flask. Plug the condenser vacuum take-off exit opposite the receiving flask to eliminate path competition and direct all distillate to the chilled collection flask. SJ 35/25 adapter fits standard Büchi rotary evaporator glassware.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)