Documentos
Z239089
Pipette fillers
red silicone bulb type
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Materiais
red silicone bulb
fabricante/nome comercial
SILRED-001
Descrição geral
Valved, choice of natural or silicone rubber bulb. Allows safe manual control of filling and discharging of liquids by finger pressure.
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 3
1 of 3
Bel-Bulb™ pipettor vinyl bulb, fits 6-8 mm pipettes, L × diam. 70 mm × 5 mm
Z136085
Bel-Bulb pipettor
Pipette bulb capacity 2 mL
Sigma-Aldrich
Z136042
Pipette bulb