Documentos Principais
Z136050
Pipette bulb
capacity 8 mL
Selecione um tamanho
3 EA
R$ 163,00
R$ 163,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
3 EA
R$ 163,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
R$ 163,00
Check Cart for Availability
Materiais
natural rubber
oval red
embalagem
pkg of 3 ea
fabricante/nome comercial
200-8
capacidade
8 mL
Categorias relacionadas
Descrição geral
Natural rubber dropper bulbs for use with pipettes ensuring precise transfer and measurement of liquids.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 8
1 of 8
Pipette bulb capacity 2 mL
Sigma-Aldrich
Z136042
Pipette bulb
Pipette bulb volume 3 mL
Sigma-Aldrich
Z509043
Pipette bulb
Pipette fillers red natural rubber bulb type
Z136468
Pipette fillers
Pasteur pipettes short capillary tip, approx 2 mL withdraw volume, soda-lime glass
Z627992
Pasteur pipettes
Questions
1–2 of 2 Questions  
  1. What are the dimensions for the 8ml pipette bulb (product Z136050)?
    1 answer
    1. The 8ml pipette bulb (product Z136050) is a natural rubber bulb with a 8ml capcity. It has the following dimensions:Bore Size: 4mmDiameter:  25mmLength:  40mm
      Helpful?
  2. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.