Documentos Principais
TEM-LC200-NI-150
Sigma-Aldrich
Lacey Carbon Supported Nickel Grid
size 200 mesh, hole size 150 μm , Average, box of 25
About This Item
Código UNSPSC:
41111761
embalagem
box of 25
tamanho do furo
150 μm , Average
tamanho
200 mesh
Descrição geral
Lacey Carbon Supported Grid: Nickel Grid, 200 mesh. Average hole size is 150 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Escolha uma das versões mais recentes:
Certificados de análise (COA)