Documentos Principais
DWK62113D-312
Kimble® 33 Expansion Tubular Borosilicate Glass Serum Vial Without Closure
Selecione um tamanho
1 PKG
R$ 9.170,00
R$ 9.170,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 PKG
R$ 9.170,00
About This Item
Código UNSPSC:
41122001
NACRES:
NB.43
R$ 9.170,00
Check Cart for Availability
Materiais
Borosilicate glass/PTFE tube
tubulated bottom
Características
closure type: no
embalagem
pkg of 1 packs
fabricante/nome comercial
Kimble® 62113D-312
Descrição geral
Autoclavable serum vials are ideally suited for the handling, containment, and storage of a variety of liquids, including reagents, culture media, chromatography samples, and more.
Aplicação
Storage and transport of samples, lyophilization, contain harsh reagents or liquids.
Características e benefícios
  • Special design provides extra strength for freeze-drying applications
  • Serum vials are lighter in weight than molded bottles, with more closely held tolerances and uniformity of glass
  • Special blowback design provides ease of use for lyophillization stoppers
  • Aluminum seals, stoppers and septa are available separately
  • Converted from Type I borosilicate glass tubing, vials conform to the hydrolytic resistance requirements outlined in USP and EP 3.2.1
Informações legais
KIMBLE is a registered trademark of DWK Life Sciences
Escolha uma das versões mais recentes:
Certificados de análise (COA)