Documentos Principais
CLS69472L
Pyrex® resin reaction kettles
capacity 2,000 mL
Sinônimo(s):
pyrex flask, pyrex with lid, pyrex with lids, Corning® 6947
Selecione um tamanho
1 EA
R$ 3.175,00
R$ 3.175,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 3.175,00
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
R$ 3.175,00
Check Cart for Availability
embalagem
case of 1 ea
fabricante/nome comercial
Corning 6947-2L
disponibilidade
not available in EU
capacidade
2,000 mL
Descrição geral
Designed for viscous reaction materials. 500 and 1,000 mL kettles have interchangeable covers with four openings (24/40 joints). 2,000, 3,000 and 4,000 mL kettles have three openings with 24/40 joints and one with a 34/45 joint. Flanges of covers and bottoms are finely ground for a tight seal. Order clamp separately.
Informações legais
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)