Documentos
CLS5341500
Pyrex® heavy wall filtering flasks
capacity 500 mL
Sinônimo(s):
conical flasks, filtering flask, filtering flasks, flasks erlenmeyer
About This Item
Código UNSPSC:
41121804
Materiais
borosilicate glass
flat bottom flask
narrow-mouth
tamanho da tampa
7
esterilidade
non-sterile
Características
closure type Rubber stopper
embalagem
case of 1
fabricante/nome comercial
Corning 5341-500
capacidade
500 mL
Descrição geral
Permanent, white enamel graduations and marking spots and replaceable PP tubulation with neoprene grommet. Tubulation is 12.5 mm o.d.
Informações legais
PYREX is a registered trademark of Corning, Inc.