Principaux documents
Z740158
LifeSep magnetic separator unit
96P, for 96 well tray, deep well
Synonyme(s) :
cell sorting magnetic seperation unit
1 EA
782,00 €
782,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
782,00 €
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NA.56
782,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
deep well
for 96 well tray
Fabricant/nom de marque
Dexter Magnetic Technologies 2501005
Catégories apparentées
Description générale
Durable, easy-to-use and designed to accommodate industry standard microtitre plates. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
Caractéristiques et avantages
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Optimized field gradient within wells
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
Les clients ont également consulté
Questions
  1. Does the magent allow for seperation on the wall of wells in comaprison to Z740159, which I asume immobilizes on the bottom? Which 96well microplates with U- or V-bottom are compatible? Whats the minimum plate height compatibility?
    1 answer
    1. The following questions would be best answered by the manufacture. The manufacture is Dexter Magnetic Technologies. The supplier product number is 2501005. See the link below to the supplier data sheet for this product:
      https://www.dextermag.com/wp-content/uploads/2017/08/LifeSep-96P.pdf
      For Product Z740159, the manufacturer part number is 2501008. See the supplier data sheet at the link below for comparison to supplier product 2501005:
      https://dextermag.eu/wp-content/uploads/2019/01/LifeSep_Biomagnetic-Separator_96F_Tray-2.pdf