Principaux documents
Z741773
LifeSep 125SX
for 125 ml vessels
Synonyme(s) :
LifeSep magnetic separation stand
1 EA
10 040,00 €
10 040,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
10 040,00 €
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NA.56
10 040,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Fabricant/nom de marque
Dexter Magnetic Technologies 2906001-1
H × l × P
71 mm × 98 mm × 72 mm
Description générale
Magnetic separators for single tubes are durable and easy-to-use. For use in cell sorting, RNA and DNA isolation and purification of biomolecules. Fast and easy aspiration is achieved with our Neodymium-Iron-Boron (Nd-Fe-B) magnet configuration.
  • Patented Nd-Fe-B magnet configuration for optimal results
  • Open-faced design for clear visibility
  • Rapid separation of micro- and nanometer beads
  • Easy aspiration
  • Superior particle retention
  • Removable base for easy cleaning
  • Durable and easy to use