Principaux documents
Z290440
Sigma-Aldrich
Sigma-Aldrich® vacuum filtration assembly
for 47 mm filters with glass support, #8 stopper, same as Z290432 but without filtration flask
Synonyme(s) :
Filtration Assembly, Vacuum Filtration Setup
1 EA
299,00 €
299,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
299,00 €
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
299,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
glass housing
glass membrane
Caractéristiques
graduation
Conditionnement
pkg of 1 ea
Diam. du col
41 mm
Description générale
Same as Z290432 but without filtration flask.
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Informations légales
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany