Z683213
J. Young high-vacuum PTFE valve, straight through
stopcock size 15 mm, side arm O.D. 19 mm
121,00 €
About This Item
121,00 €
Produits recommandés
Fabricant/nom de marque
J. Young
D.E.du bras latéral
19 mm
Taille du robinet d'arrêt
15 mm
Description générale
Features:
- PTFE piston with unique PTFE-sheathed elastomer O-ring seals
- Grease-free glass-to-PTFE seals
- High-vacuum to 10-6 torr