This product is a standard GL45 hole cap with an insert that accommodates two ports. The cap hole diameter is 1.4 inches or 35.5 mm.
Z543845
VerSA-Flow™ delivery cap
two-hole
228,00 €
About This Item
228,00 €
Produits recommandés
Matériaux
PBT
D.E. du tuyau
1/8 in.
Raccords
GL 45
Contenu apparenté
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.
-
Hi I'm just wondering about : VerSA-Flow™ delivery cap two-hole Z543845 What it's Inner diameter