Principaux documents
Z408484
Sigma-Aldrich
Floating CryoTube® rack
polypropylene, autoclavable
1 EA
37,00 €
37,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
37,00 €
About This Item
Code UNSPSC :
41122806
Nomenclature NACRES :
NB.22
37,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polypropylene
Caractéristiques
autoclavable
Holds 20 x cryotubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art F18876-0020
Technique(s)
cryopreservation: suitable
Description générale
Round, rack holds 20 cryotubes tightly in a circular pattern formed by 12 outer and 8 inner openings. Excellent for holding cryotubes during work or incubation; rack floats in water and can also be used for quickly thawing frozen cells in a 37 °C water bath. The filled rack unloads easily by pushing the rack down against the bench until the legs touch, releasing the tubes.
33/4 diam. × 1/4 in. thick with 3/4 in. legs.
Informations légales
CryoTube is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Les clients ont également consulté
Questions
  1. What is the diameter of the holes?
    1 answer
    1. The holes present in this floating rack have a diameter of 0.498 inches (12.65 mm).