Principaux documents
Z267147
Sartorius balance analytical
Model BP300S, AC/DC input 220 V AC
About This Item
Code UNSPSC :
41111502
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Fabricant/nom de marque
Model BP300S
Description générale
  • Large LCD readout
  • Push button internal calibration
  • Large, glass draft chamber
  • ISO/GLP compliance recording
  • RS232 serial interface
  • Overload protection
  • Level indicator
  • Lug for antitheft locking
  • CE mark of conformity
The industry standard for weighing tasks requiring fast, accurate results. Fully automatic calibration on all models ensures results are perfectly accurate with the press of a key. Convenient application programs offer parts counting, weighing in %, net total formulation, and mass unit conversions.