Documents
Z564907
Buchi® diagonal condenser, for Rotavapor® evaporators
for R210/215, glass
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
glass
Fabricant/nom de marque
Büchi 46988
Description générale
Condensers for R200/205 and R210/215 come with hose barbs and drain sleeve. Replacement condensers do not include stopcocks.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG
Rotavapor is a registered trademark of Büchi Labortechnik AG