Principaux documents
Z190284
Arrow electric lab stirrer
speed 0-6000 rpm, direct, output0.10 hp
Synonyme(s) :
lab mixer, lab mixers, mixer stirrer
1 EA
964,00 €
964,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
964,00 €
About This Item
Code UNSPSC :
41103801
Nomenclature NACRES :
NB.72
964,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Entrée CA/CC
115 V
115 V
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Arrow Engineering 6000
Disponibilité
not available in EU
Paramètres
0-6000 rpm speed
Sortie
0.10 hp
Description générale
Totally enclosed, non-ventilated motor seals out corrosive fumes and dust. With on/off switch, circuit-breaker, and variable speed control. Includes a 9/16 × 10 in. support rod and 3/8 diam. × 12 in. L SS shaft and propeller.
Qualité
Note: Arrow lab stirrers conform to CSA22.2 but are not certified
Questions
  1. Is this stirrer fitted with blades?
    1 answer
    1. This mixer includes a 2.5 inch propeller mounted on a 12 inch shaft. The shaft diameter is 3/8 inches. The propeller image is provided below.