Principaux documents
CAD6019
1-way stopcock
female Luer lock to male Luer
Synonyme(s) :
Stopcock Valve
1 EA
96,80 €
96,80 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
96,80 €
About This Item
Code UNSPSC :
41122000
Nomenclature NACRES :
NB.03
96,80 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Cadence Science 6019
Catégories apparentées
Description générale
Stopcocks are precision-machined, individually lapped, and carefully mated to maintain pressure characteristics. These stopcocks are easily integrated with other stopcocks, manifolds, adapters, tubing, needles, and glass syringes.
  • Made from nickel plated brass or stainless steel
  • Tested to 45 psi water pressure
  • High pressure stopcocks are rated to 290 psi