Skip to Content
Merck
All Photos(1)

Key Documents

R4503

Sigma-Aldrich

Sma I from Serratia marcescens Sb

Restriction Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

grade

for molecular biology

form

buffered aqueous glycerol solution

concentration

10,000 units/mL

shipped in

wet ice

storage temp.

−20°C

Specificity

Recognition sequence: 5′-CCC/GGG-3′
Cutting results: a 2-10-fold Sma I overdigestion of 1 μg λ DNA substrate results in 100% cutting
Heat inactivation: 65 °C for 15 minutes.

Application

SmaI is a restriction endonuclease used to cut DNA at the recognition sequence 5′-CCC/GGG-3′, generating DNA fragments with blunt termini.

Other Notes

Supplied with 10x Restriction Enzyme Buffer SA (B7531).
Comment: Half-life of Sma I at 37 °C is approximately 15 minutes.

Linkage

Isoschizomer: Xma I. Unlike Xma I, Sma I produces blunt-ended fragments.

Unit Definition

One unit is the enzyme activity that completely cleaves 1 mg of λDNA in 1 hr. at 25 °C in a total volume of 25 ml of 1x digestion buffer SA forrestriction enzymes.

Physical form

Solution in 10 mM potassium phosphate, pH 7.0, 1 mM EDTA, 300 mM NaCl, 5 mM dithiothreitol, 50% glycerol (v/v) , 0.02% polydocanol (v/v), at 4 °C

incubation buffer

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leila C Campos et al.
Diagnostic microbiology and infectious disease, 65(2), 112-115 (2009-08-28)
The newly described Streptococcus pneumoniae serotype 6C accounted for 2.3% (16/709) of meningitis cases and 3.2% (3/95) of nasopharyngeal isolates from healthy individuals in Brazil. The strains were multidrug resistant (18.8%) and genetically diverse. Despite low serotype 6C prevalence, continuous
D Savoia et al.
The Journal of antimicrobial chemotherapy, 45(1), 41-47 (2000-01-11)
Two hundred and twenty one Streptococcus pyogenes isolates collected from throat swabs of untreated children with uncomplicated pharyngotonsillitis living in two centres situated in the north of Italy were tested to evaluate their macrolide resistance phenotype. Isolates were also typed
Two restriction-like enzymes from Xanthomonas malvacearum.
S A Endow et al.
Journal of molecular biology, 112(3), 521-529 (1977-05-25)
C Kessler et al.
Gene, 92(1-2), 1-248 (1990-08-16)
The properties and sources of all known class-I, class-II and class-III restriction endonucleases (ENases) and DNA modification methyltransferases (MTases) are listed and newly subclassified according to their sequence specificity. In addition, the enzymes are distinguished in a novel manner according
Saqib H Ansari et al.
Journal of pediatric hematology/oncology, 35(4), e153-e156 (2013-02-08)
β-thalassemia is characterized by impaired β-chain synthesis leading to ineffective erythropoiesis, severe anemia, and a need for blood transfusion. Presence of Xmn I polymorphism (-158 C-T nucleotide change) in γ-globin gene is associated with a higher fetal hemoglobin and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service