Merck
All Photos(4)
Key Documents
SQPAKQUAM1
Milli-Q
SQPAK Quanta Cartridge
greener alternative
Produces ultrapure (Type 1) water for use on the ultrapure dispensing module of Milli-Q® SQ 2Series systems
Select a Size
1 EACH
€346.00
€346.00
Available to ship on15 April 2025Details
Request more information
Select a Size
Change View
1 EACH
€346.00
About This Item
UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
NB.35
€346.00
Available to ship on15 April 2025Details
Request more information
Quality Level
packaging
pkg of 1 unit
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
compatibility
for use with Milli-Q® SQ 2Series
greener alternative category
General description
To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
SQPAK Quanta cartridge is a high-capacity polishing cartridge that produces ultrapure (Type 1) water at 18.2 MΩ·cm resistivity.
The cartridge contains IQnano ion-exchange media to remove ions down to trace levels.
For use on the dispensing module of Milli-Q® SQ 2Series configurations, such as Milli-Q® SQ 200/200P and Milli-Q® SQ 240C/240CV systems.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
Features and Benefits
  • Easy replacement thanks to the Twist & Lock mechanism
  • Need for cartridge replacement is indicated by an orange blinking pictogram light on the system
  • Ultrapure label and blue color of the SQPAK Quanta cartridge are repeated on the production station interface, to inform about its water purification purpose and guide replacement
Legal Information
IQNANO is a trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SQPAK is a trademark of Merck KGaA, Darmstadt, Germany
Visit the Document Library
Protocols
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
Related Content
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.