AXYTF20LRS
MAXYMum Recovery® pipette filter tips
for P-20, racked, size 20 μL, pkg of 5x960tips/cs, sterile
Synonym(s):
Pipette tips, pipette tips
Select a Size
€624.00
Select a Size
About This Item
RNase free
barrier: no
endotoxin free
filter
Corning
€624.00
Recommended Products
material
clear
sterility
sterile
feature
DNA free
RNase free
barrier: no
endotoxin free
filter
manufacturer/tradename
Axygen Inc TF-20-L-R-S
Corning
packaging
pkg of 5x960tips/cs
size
20 μL
volume
20 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Features and Benefits
- Tips create a physical barrier to prevent cross-contamination and block aerosols
- The DNA inhibitors free-polyethylene filters are fit into the tips robotically to prevent nuclease contamination
- Packed in sealed-bottom racks
- Tips are RNAse and DNAse free
- Non-pyrogenic
- High accuracy, yield, and precision
- Sample loss is reduced due to high fluid retention
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service