Skip to Content
Merck
All Photos(1)

Key Documents

AXYT300L

Axygen® Maxymum Recovery® Microvolume Pipet Tips

for P2/P10, size 0.5-10 μL, pkg of 20x1000tips/cs, clear

Synonym(s):

MAXYMum Recovery® pipette tips, Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€684.00

€684.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€684.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
0.5—10 μL
sterility:
non-sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 20x1000tips/cs
manufacturer/tradename:
Axygen Inc T-300-L
Corning

€684.00


Please contact Customer Service for Availability

material

clear

sterility

non-sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc T-300-L
Corning

packaging

pkg of 20x1000tips/cs

size

0.5-10 μL

volume

0.5—10 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Related Categories

Features and Benefits

Axygen® MAXYMum Recovery® pipette tips provide the following benefits:
  • Tips fit a broad range of single and multi-channel pipettors
  • Compatible with most of the well-known brands of pipettors
  • Tips are made of polypropylene and are non-pyrogenic
  • Tips are RNAse and DNAse free
  • Flexible walls and a series of internal sealing rings provides good fitting which requires less force to load and eject
  • Microvolume
  • Low-risk of sample loss due to high fluid retention
  • Low-risk of cross-contamination
  • Autoclavable
  • Higher yield, accuracy, and precision

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Maxymum Recovery is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service