Skip to Content
Merck
All Photos(1)

Key Documents

AXYT300LRS

MAXYMum Recovery® pipette tips

for P2/P10, racked, size 0.5-10 μL, pkg of 5x960tips/cs, clear, sterile

Synonym(s):

Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€504.00

€504.00


Please contact Customer Service for Availability


Select a Size

Change View
1 PKG
€504.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.02
volume:
0.5—10 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter: no
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc T-300-L-R-S
Corning

€504.00


Please contact Customer Service for Availability

material

clear

sterility

sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter: no

manufacturer/tradename

Axygen Inc T-300-L-R-S
Corning

packaging

pkg of 5x960tips/cs

size

0.5-10 μL

volume

0.5—10 μL

color

clear

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

  • Corning® Maxymum Recovery® microvolume pipette tips are made of polypropylene.
  • The tips are optimized for use with both single and multi-channel pipettors.
  • Features a flexible wall and internal sealing ring for a secure fit.
  • Maxymum Recovery technology for ultra-low retention, excellent precision, accuracy, and high yield.

Other Notes

Browse all pipette tips

Legal Information

Corning is a registered trademark of Corning, Inc.
Maxymum Recovery is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service