Skip to Content
Merck
All Photos(1)

Key Documents

AXYTF300RS

Pipette filter tips

for P-2, ultramicro, racked, size 0.5-10 μL, pkg of 5x960tips/cs, sterile

Synonym(s):

Pipette tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume:
0.5—10 μL
sterility:
sterile
feature:
DNA free
RNase free
barrier: no
endotoxin free
filter
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc TF-300-R-S
Corning
Pricing and availability is not currently available.

material

clear

sterility

sterile

feature

DNA free
RNase free
barrier: no
endotoxin free
filter

manufacturer/tradename

Axygen Inc TF-300-R-S
Corning

packaging

pkg of 5x960tips/cs

size

0.5-10 μL

volume

0.5—10 μL

Looking for similar products? Visit Product Comparison Guide

Application

Axygen® Filter Barrier tips can be used for microbiological and DNA amplification studies where contamination can tamper the results.

Features and Benefits

Axygen® Filter Barrier tips have the following features and benefits:
  • Tips create a physical barrier to prevent cross-contamination and block aerosols
  • The DNA inhibitors free-polyethylene filters are fit into the tips robotically to avoid nuclease contamination
  • Packed in sealed-bottom racks
  • Tips are RNAse and DNAse free
  • Non-pyrogenic

Other Notes

Browse all pipette tips

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service