SYMBLFH01CN
Steritest® Symbio LFH pump kit
Bench-top usage in laminar flow hood; 2 media, with power cord for China
Synonym(s):
Steritest® Symbio LFH Pump Kit, Peristaltic pump; Sterility Testing pump
Select a Size
€21,560.00
Estimated to ship on20 May 2025Details
Select a Size
About This Item
€21,560.00
Estimated to ship on20 May 2025Details
Recommended Products
AC/DC input
100 V / 240 V, 50 -60 Hz
Quality Level
material
316L stainless steel (Pump, Pump Head Cover, Bottle Holder, Drain Tray Support)
EPDM rubber seal
polyester (Keypad)
polyphenylsulfone (PPSU) (Tubing Guide, Drain Tray Container)
silicone seal
window(s) (toughened glass)
Agency
EP
JP
USP
manufacturer/tradename
Steritest®
parameter
15.8 kg operating weight (with accessories)
pump H × W
41.0 cm × 63.3 cm , with accessories
pump body H × D
28.2 cm × 37.2 cm , with accessories
pump head H
15.8 cm
application(s)
pharmaceutical
sterility testing
(Industrial microbiology)
compatibility
Steritest® devices
Looking for similar products? Visit Product Comparison Guide
General description
The automatic closure eliminates the risk of glove pinching and ensures reliable splitting of the liquid sample. The drain tray and bottle holder are ergonomically positioned around the pump head for easy access in limited-space environments. Furthermore, the drain tray is designed to minimize splashing under the canister base. Two pressure detection modes (including automatic pumping speed reduction) alert the operator, reducing the risk of sterility test interruption and minimizing the stress on microorganisms.
A power cord specific for China is available with this pump. A wide range of optional accessories are also offered to improve the sterility testing workflow.
Application
Features and Benefits
- Easy-to-use design
- Reliable
- Safe
- Ergonomic
- User-friendly
Components
- Peristaltic pump
- Guides quality certificate
- Bottle holder
- Drain tray
- Silicone drain tubing
- Communication hub
- Power cord
- Network cable
- USB flash drive
- 3 quick guides
Legal Information
accessory
also commonly purchased with this product
recommended
related product
required but not provided
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service