Skip to Content
Merck
All Photos(1)

Key Documents

SYMBDTH01

Millipore

Steritest® Symbio complete drain tray

For 2 canisters, LFH pump

Synonym(s):

Outlet

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€1,460.00

€1,460.00


Estimated to ship on15 April 2025Details


Request more information

Select a Size

Change View
1 UNIT
€1,460.00

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

€1,460.00


Estimated to ship on15 April 2025Details


Request more information

material

316 stainless steel container (drain tray)
EPDM rubber seal

Agency

EP (2.6.1)
JP (4.06)
USP (71)

packaging

pkg of 1 unit

manufacturer/tradename

Steritest®

input

pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio pump

shipped in

ambient

General description

Steritest® Symbio complete drain tray includes stainless steel canister holder, plastic fluid collector, and seal. This tray is designed for 2 canisters LFH Pump. The Steritest® Symbio LFH Pump is designed for different testing environments, including a laminar flow hood, biosafety cabinet, cleanroom, or even inside an isolator.

Application

Steritest® Symbio complete drain tray is a Steritest® Symbio LFH Pump accessory for streamlining your workflow and increasing safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service